Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetylcorynoline | 18797-80-3 | 409.43 | 5 MG
SDP

Catalog No. 5000416798
Encompass_Preferred

Acetylcorynoline is the major alkaloid component derived from *Corydalis bungeana* and exhibits anti-inflammatory properties. It significantly inhibits the secretion of tumor necrosis factor-α, interleukin-6, and interleukin-12p70 by LPS-stimulated Dendritic cells (DCs), and inhibits LPS-induced activation of IκB kinase and mitogen-activated protein kinase.

  • Purity: 99.70%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Plants (other families)
  • Structure classification: Alkaloids, isoquinoline alkaloids
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-004-16798 Supplier Medchemexpress LLC Supplier No. HYN07595MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetylcorynoline is the major alkaloid component derived from *Corydalis bungeana* and exhibits anti-inflammatory properties. It significantly inhibits the secretion of tumor necrosis factor-α, interleukin-6, and interleukin-12p70 by LPS-stimulated Dendritic cells (DCs), and inhibits LPS-induced activation of IκB kinase and mitogen-activated protein kinase.

  • Purity: 99.70%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Plants (other families)
  • Structure classification: Alkaloids, isoquinoline alkaloids
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.