Learn More
TARGETMOL CHEMICALS INC Acetylcorynoline 20MG
SDP
Catalog No. 502880027
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. 1. Acetylcorynoline (O-Acetylcorynoline) has antifungal、anti-inflammatory activity. 2. Acetylcorynoline may be one of the potent immunosuppressive agents through the blockage of dendritic cells maturation and function. 3. Acetylcorynoline is potential as a possible antiparkinsonian drug. (1) Significantly decreases dopaminergic neuron degeneration induced by 6-hydroxydopamine in BZ555 strain; (2) Prevents alpha-synuclein aggregation; (3) Restores food-sensing behavior, and dopamine levels; (4) Prolongs life-span in 6-hydroxydopamine-treated N2 strain. 4. Acetylcorynoline may exert its effects by decreasing egl-1 expression to suppress apoptosis pathways and by increasing rpn5 expression to enhance the activity of proteasomes. Purity 99.45%
Catalog No. 50-288-0027 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S235720MG
Add to Cart
Add to Cart
Also available in 1 mL and bulk. Please contact Fisher for quotes. 1. Acetylcorynoline (O-Acetylcorynoline) has antifungal、anti-inflammatory activity. 2. Acetylcorynoline may be one of the potent immunosuppressive agents through the blockage of dendritic cells maturation and function. 3. Acetylcorynoline is potential as a possible antiparkinsonian drug. (1) Significantly decreases dopaminergic neuron degeneration induced by 6-hydroxydopamine in BZ555 strain; (2) Prevents alpha-synuclein aggregation; (3) Restores food-sensing behavior, and dopamine levels; (4) Prolongs life-span in 6-hydroxydopamine-treated N2 strain. 4. Acetylcorynoline may exert its effects by decreasing egl-1 expression to suppress apoptosis pathways and by increasing rpn5 expression to enhance the activity of proteasomes. Purity 99.45%