Learn More
Medchemexpress LLC HY-B0215 10g , Acetylcysteine N-Acetyl-L-cysteine;LNAC;NAC CAS:616-91-1 Purity:98%
SDP
Catalog No. 501492151
Encompass_Preferred
Medchemexpress, HY-B0215 10g Acetylcysteine N-Acetyl-L-cysteine;LNAC;NAC CAS:616-91-1 Formula:C5H9NO3S Purity:98% Acetylcysteine is used mainly as a mucolytic, which protects against acetaminophen overdose-induced hepatotoxicity by maintaining or restoring hepatic concentrations of glutathione. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2151 Supplier Medchemexpress LLC Supplier No. HYB021510G
Medchemexpress, HY-B0215 10g Acetylcysteine N-Acetyl-L-cysteine;LNAC;NAC CAS:616-91-1 Formula:C5H9NO3S Purity:98% Acetylcysteine is used mainly as a mucolytic, which protects against acetaminophen overdose-induced hepatotoxicity by maintaining or restoring hepatic concentrations of glutathione. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.