Learn More
U.S. Pharmacopeia Acetylcysteine, 616-91-1, MFCD00004880, 200 mg
Catalog No. NC1447009
Encompass
Linear Formula: HSCH2CH(NHCOCH3)CO2H, Molecular Weight: 163.19, mp: 106-108 °C, Synonym: N-Acetyl-L-cysteine, LNAC, NAC.
Catalog No. NC1447009 Supplier U.S. Pharmacopeia Supplier No. 1009005
Linear Formula: HSCH2CH(NHCOCH3)CO2H, Molecular Weight: 163.19, mp: 106-108 °C, Synonym: N-Acetyl-L-cysteine, LNAC, NAC.