Learn More
Medchemexpress LLC HY-B0215 500mg , Acetylcysteine CAS:616-91-1 Purity:>98%
SDP
Catalog No. 502259210
Encompass_Preferred
HY-B0215 500mg Acetylcysteine CAS: 616-91-1 Acetylcysteine (N-Acetylcysteine) is a mucolytic agent which reduces the thickness of the mucus. Acetylcysteine is a ROS inhibitor. Acetylcysteine is a cysteine precursor, prevents hemin-induced ferroptosis by neutralizing toxic lipids generated by arachidonate-dependent activity of 5-lipoxygenases[5]. Acetylcysteine induces cell apoptosis. Acetylcysteine also has anti-influenza virus activities[7]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9210 Supplier Medchemexpress LLC Supplier No. HYB0215500MG
HY-B0215 500mg Acetylcysteine CAS: 616-91-1 Acetylcysteine (N-Acetylcysteine) is a mucolytic agent which reduces the thickness of the mucus. Acetylcysteine is a ROS inhibitor. Acetylcysteine is a cysteine precursor, prevents hemin-induced ferroptosis by neutralizing toxic lipids generated by arachidonate-dependent activity of 5-lipoxygenases[5]. Acetylcysteine induces cell apoptosis. Acetylcysteine also has anti-influenza virus activities[7]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.