Learn More
Medchemexpress LLC Acetylcysteine (Standard) | 616-91-1 | 98.2% | 25 MG
SDP
Catalog No. 5000451029
Encompass_Preferred
Acetylcysteine (Standard) is an analytical standard of Acetylcysteine, designed for research and analytical applications. It functions as a mucolytic agent by reducing mucus thickness, acts as a ROS inhibitor, and serves as a cysteine precursor. This compound also prevents hemin-induced ferroptosis, induces cell apoptosis, and possesses anti-influenza virus activities. As the most stable form of cysteine for drug delivery, it can be utilized in disulfidptosis studies.
  • Reduces mucus thickness
  • Acts as a ROS inhibitor and cysteine precursor
  • Prevents hemin-induced ferroptosis
  • Induces cell apoptosis
  • Possesses anti-influenza virus activities
  • Utilized in disulfidptosis studies
  • Suitable for qualitative, quantitative, and methodological research experiments
Catalog No. 50-004-51029 Supplier Medchemexpress LLC Supplier No. 25MGHYB0215R
Acetylcysteine (Standard) is an analytical standard of Acetylcysteine, designed for research and analytical applications. It functions as a mucolytic agent by reducing mucus thickness, acts as a ROS inhibitor, and serves as a cysteine precursor. This compound also prevents hemin-induced ferroptosis, induces cell apoptosis, and possesses anti-influenza virus activities. As the most stable form of cysteine for drug delivery, it can be utilized in disulfidptosis studies.
  • Reduces mucus thickness
  • Acts as a ROS inhibitor and cysteine precursor
  • Prevents hemin-induced ferroptosis
  • Induces cell apoptosis
  • Possesses anti-influenza virus activities
  • Utilized in disulfidptosis studies
  • Suitable for qualitative, quantitative, and methodological research experiments