Learn More
Cayman Chemical AcetylDLcarnItInchlorIde 100mg
SDP
Catalog No. 502988387
Encompass_Preferred
A racemic mixture of acetyl-D-carnitine and acetyl-L-carnitine; increases the oxidation rate of the branched-chain 2-oxo acid 3-methyl-2-butanoate in isolated rat muscle mitochondria in the absence of carnitine at 1 mM; decreases the hind limb vasculature blood flow rate in rats; decreases the levels of palmitic acid and oleic acid in the cerebral cortex during the recovery period in a dog model of insulin-induced hypoglycemic brain injury
Catalog No. 50-298-8387 Supplier Cayman Chemical Supplier No. 31015100 MG
A racemic mixture of acetyl-D-carnitine and acetyl-L-carnitine; increases the oxidation rate of the branched-chain 2-oxo acid 3-methyl-2-butanoate in isolated rat muscle mitochondria in the absence of carnitine at 1 mM; decreases the hind limb vasculature blood flow rate in rats; decreases the levels of palmitic acid and oleic acid in the cerebral cortex during the recovery period in a dog model of insulin-induced hypoglycemic brain injury