Learn More
Cayman Chemical Acetyl-DL-carnitine (chloride), Purity: ≥98%, CAS: 2504-11-2, 50 mg
SDP
Catalog No. 502989500
Encompass_Preferred
A racemic mixture of acetyl-D-carnitine and acetyl-L-carnitine; increases the oxidation rate of the branched-chain 2-oxo acid 3-methyl-2-butanoate in isolated rat muscle mitochondria in the absence of carnitine at 1 mM; decreases the hind limb vasculature blood flow rate in rats; decreases the levels of palmitic acid and oleic acid in the cerebral cortex during the recovery period in a dog model of insulin-induced hypoglycemic brain injury
Catalog No. 50-298-9500 Supplier Cayman Chemical Supplier No. 3101550 MG
A racemic mixture of acetyl-D-carnitine and acetyl-L-carnitine; increases the oxidation rate of the branched-chain 2-oxo acid 3-methyl-2-butanoate in isolated rat muscle mitochondria in the absence of carnitine at 1 mM; decreases the hind limb vasculature blood flow rate in rats; decreases the levels of palmitic acid and oleic acid in the cerebral cortex during the recovery period in a dog model of insulin-induced hypoglycemic brain injury