Learn More
Strem, An Ascensus Company CAS# 1333-86-4. 1kg. Acetylene carbon black (100% compressed). MFCD00133992
SDP
Catalog No. 5090108776
Encompass
CAS# 1333-86-4. 1kg. Acetylene carbon black (100% compressed). MFCD00133992. Molecular Weight: 12.011. Molecular Formula: C. Color/form: black pwdr. (avg. particle size 0.042 microns) Strem# 06-0025. http://www.strem.com/catalog/v/06-0025/
Catalog No. 50-901-08776 Supplier Strem, An Ascensus Company Supplier No. 0600251000G
CAS# 1333-86-4. 1kg. Acetylene carbon black (100% compressed). MFCD00133992. Molecular Weight: 12.011. Molecular Formula: C. Color/form: black pwdr. (avg. particle size 0.042 microns) Strem# 06-0025. http://www.strem.com/catalog/v/06-0025/