Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylene-PEG4-biotin conjugate, Thermo Scientific™

Catalog No. AAJ65577MC
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65577MC 100 mg
AAJ65577MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65577MC Supplier Thermo Scientific Chemicals Supplier No. J65577MC

An acetylene-biotin conjugate with PEG spacer

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1262681-31-1
Molecular Formula C21H35N3O6S
Molecular Weight (g/mol) 457.59
MDL Number MFCD22380755
InChI Key SKMJWNZZFUDLKQ-UHFFFAOYNA-N
Synonym biotin-peg4-alkyne, acetylene-peg4-biotin conjugate
PubChem CID 75409985

Specifications

Color White
Quantity 100 mg
Formula Weight 457.59
Physical Form Solid
Chemical Name or Material Acetylene-PEG4-biotin conjugate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.