Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyleugenol, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111001822 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1822 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1822 Supplier MilliporeSigma Supelco Supplier No. 36103100MG

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content incl. uncertainty and expiry date are given on the certificate.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 93-28-7
Density 1.079 g/mL (at 25°C)
Flash Point 110°C - Closed Cup
Shipping Condition Wet ice
Linear Formula C12H14O3
MDL Number MFCD00026191
Boiling Point 281°C to 286°C
Synonym Eugenyl acetate; 4-Allyl-2-methoxyphenyl acetate; O-Acetyleugenol; Eugenol acetate
Type Reference Material
Quantity 100 mg
Formula Weight 206.24
Percent Purity ≥98.0% (GC)
Grade Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.