Learn More
Cayman Chemical Acetylharpagide, Purity: ≥98%, CAS: 6926-14-3, 50 mg
SDP
Catalog No. 502942639
Encompass_Preferred
An iridoid glycoside with diverse biological activities; reduces the proliferation of A431 cancer cells (IC50 = 310 µM); reduces the number of tumors per mouse and the percentage of mice with tumors in a mouse model of 4-nitroquinolin 1-oxide-induced carcinogenesis at 0.005% in the drinking water; decreases carrageenan-induced paw swelling in rats at 10 mg/kg; reduces blood glucose levels in a mouse model of alloxan-induced type I diabetes at 10 mg/kg
Catalog No. 50-294-2639 Supplier Cayman Chemical Supplier No. 3993050 MG
An iridoid glycoside with diverse biological activities; reduces the proliferation of A431 cancer cells (IC50 = 310 µM); reduces the number of tumors per mouse and the percentage of mice with tumors in a mouse model of 4-nitroquinolin 1-oxide-induced carcinogenesis at 0.005% in the drinking water; decreases carrageenan-induced paw swelling in rats at 10 mg/kg; reduces blood glucose levels in a mouse model of alloxan-induced type I diabetes at 10 mg/kg