Learn More
Cayman Chemical Acetyl-L-carnitine-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), CAS: 362049-62-5, 50 mg
SDP
Catalog No. 502935142
Encompass_Preferred
An internal standard for the quantification of L-acetylcarnitine by GC- or LC-MS
Catalog No. 50-293-5142 Supplier Cayman Chemical Supplier No. 2656450 MG
An internal standard for the quantification of L-acetylcarnitine by GC- or LC-MS