Learn More
Medchemexpress LLC Acetylleucine (N-acetyl-DL-leucine) | 99-15-0 | 99.4% | 5 G
SDP
Catalog No. 5000412651
Encompass_Preferred
Acetylleucine (N-Acetyl-DL-leucine) is an orally active compound that can be used for the research of acute vestibular vertigo, cerebellar ataxia, and nystagmus.
  • Orally active compound
  • Used for research of acute vestibular vertigo, cerebellar ataxia, and nystagmus
  • Accelerates compensation of postural symptoms after unilateral labyrinthectomy in rats
Catalog No. 50-004-12651 Supplier Medchemexpress LLC Supplier No. HYB14425G
Acetylleucine (N-Acetyl-DL-leucine) is an orally active compound that can be used for the research of acute vestibular vertigo, cerebellar ataxia, and nystagmus.
  • Orally active compound
  • Used for research of acute vestibular vertigo, cerebellar ataxia, and nystagmus
  • Accelerates compensation of postural symptoms after unilateral labyrinthectomy in rats