Learn More
Medchemexpress LLC HY-126237 5mg Medchemexpress, Acetyllovastatin CAS:81189-92-6 Purity:>98%
SDP
Catalog No. 501932616
Encompass_Preferred
Medchemexpress, HY-126237 5mg Acetyllovastatin CAS:81189-92-6 Acetyllovastatin, a acetate of Lovastatin, presentes a moderate inhibitory effect against the enzyme acetylcholinesterase with an IC 50 of 79 μg/mL. Lovastatin has been found to display antifungal activity, and suppresses proliferation of a number of transformed cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2616 Supplier Medchemexpress LLC Supplier No. HY1262375MG
Medchemexpress, HY-126237 5mg Acetyllovastatin CAS:81189-92-6 Acetyllovastatin, a acetate of Lovastatin, presentes a moderate inhibitory effect against the enzyme acetylcholinesterase with an IC 50 of 79 μg/mL. Lovastatin has been found to display antifungal activity, and suppresses proliferation of a number of transformed cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.