Learn More
Cayman Chemical Acetyl-β-methylcholine (chloride), Purity: ≥95%, CAS: 62-51-1, 25 g
SDP
Catalog No. 502994003
Encompass_Preferred
A muscarinic agonist; decreases heart rate and blood pressure and increases the airway pressure time index in a dose-dependent manner in wild-type mice
Catalog No. 50-299-4003 Supplier Cayman Chemical Supplier No. 2309225 G
A muscarinic agonist; decreases heart rate and blood pressure and increases the airway pressure time index in a dose-dependent manner in wild-type mice