Learn More
Cayman Chemical Acetyl-β-methylcholine (chloride), Purity: ≥95%, CAS: 62-51-1, 5 g
SDP
Catalog No. 502995541
Encompass_Preferred
A muscarinic agonist; decreases heart rate and blood pressure and increases the airway pressure time index in a dose-dependent manner in wild-type mice
Catalog No. 50-299-5541 Supplier Cayman Chemical Supplier No. 230925 G
A muscarinic agonist; decreases heart rate and blood pressure and increases the airway pressure time index in a dose-dependent manner in wild-type mice