Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetylsalicylic Acid
SCI_ED

Catalog No. S25122 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Lab Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25122 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Synonym aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
PubChem CID 2244
ChEBI CHEBI:15365
IUPAC Name 2-(acetyloxy)benzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Specifications

Density 1.35g/mL
Packaging Lab Grade
Linear Formula 2·(CH3CO2)C6H4CO2H
Quantity 100 g
Formula Weight 180.16
Physical Form Solid
Chemical Name or Material Acetylsalicylic Acid
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.