Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylsalicylic acid, 99%, Thermo Scientific Chemicals

Catalog No. AC158180500
Change view
Click to view available options
Quantity:
50 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158180500 50 g Plastic bottle
AC158185000 500 g Plastic bottle
AC158180025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158180500 Supplier Thermo Scientific Chemicals Supplier No. 158180500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Acetylsalicylic acid, aspirin, CAS# 50-78-2, is a non-steroidal anti-inflammatory that is used widely as an analgesic and antipyretic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Acetylsalicylic acid is a common non-steroidal anti-inflammatory (NSAID) and
• Acetylsalicylic acid works by inhibiting cyclooxygenases, reducing prostaglandin production and the release of pyrogens


Applications
• Acetylsalicylic acid is useful for research on fever, postoperative pain, blood clots, and inflammation
• Acetylsalicylic acid may also be used in the study of cancer prevention
• Intended for research use only

Literature references

Levine, G.N.; Bates, E.R.; Bittl, J.A.; Brindis, R.G.; Fihn, S.D.; Fleisher, L.A.; Granger, C.B.; Lange, R.A.; Mack, M.J.; Mauri, L.; Mehran, R.; Mukherjee, D.; Newby, L.K.; O'Gara, P.T.; Sabatine, M.S.; Smith, P.K.; Smith, S.C. Jr. 2016, ACC/AHA Guideline Focused Update on Duration of Dual Antiplatelet Therapy in Patients With Coronary Artery Disease: A Report of the American College of Cardiology/American Heart Association Task Force on Clinical Practice Guidelines. J Am Coll Cardiol. 2016, 68(10), 1082-115.

Hoffman, M.K.; Goudar, S.S.; Kodkany, B.S.; Metgud, M.; Somannavar, M.; Okitawutshu, J.; Lokangaka, A.; Tshefu, A.; Bose, C.L.; Mwapule, A.; Mwenechanya, M.; Chomba, E.; Carlo, W.A.; Chicuy, J.; Figueroa, L.; Garces, A.; Krebs, N.F.; Jessani, S.; Zehra F.; Saleem, S.; Goldenberg, R.L.; Kurhe, K.; Das, P.; Patel, A.; Hibberd, P.L.; Achieng, E.; Nyongesa, P.; Esamai, F.; Liechty, E.A.; Goco, N.; Hemingway-Foday, J.; Moore, J.; Nolen, T.L.; McClure, E.M.; Koso-Thomas, M.; Miodovnik, M.; Silver, R.; Derman, R.J. ASPIRIN Study Group. Low-dose aspirin for the prevention of preterm delivery in nulliparous women with a singleton pregnancy (ASPIRIN): a randomised, double-blind, placebo-controlled trial. Lancet. 2020, 25, 395(10220), 285-293.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Synonym aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
PubChem CID 2244
ChEBI CHEBI:15365
IUPAC Name 2-acetyloxybenzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 136.0°C to 140.0°C
Color White
Density 1.3500g/mL
Flash Point 250°C
Loss on Drying 0.5% max. (1 g) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula 2-(CH3CO2)C6H4CO2H
Quantity 50 g
Beilstein 10, 67
Specific Gravity 1.35
Merck Index 15, 841
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: 1g in 5mL alcohol,17mL chloroform,,and 10-15 mL ether,slightly soluble in anhydrous ether,decomposes in boiling water,alkali
Formula Weight 180.16
Percent Purity 99%
Physical Form Granular Powder
Chemical Name or Material Acetylsalicylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 200-064-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.