Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetylsalicylic acid-d3 | 921943-73-9 | MFCD09752106 | 98.5% | 183.18 g/mol | C9H5D3O4 | 5 MG
SDP

Catalog No. 5000223005
Encompass_Preferred

Aspirin-d3 (acetylsalicylic acid-d3) is a deuterium-labeled analogue of aspirin provided for laboratory research. It is used as a stable-isotope internal standard or tracer in analytical, pharmacokinetic, and biochemical studies where a labeled form of aspirin is required. This compound is for research use only and is not for human consumption or clinical applications.

  • Molecular formula C9H5D3O4.
  • Molecular weight 183.18 g/mol.
  • High reported purity (~98.5%).
  • Soluble in DMSO for convenient sample preparation.
  • Supplied in small research pack sizes such as 5 mg for method development and standards.
  • Suitable as an internal standard for LC-MS and related assays.

Catalog No. 50-002-23005 Supplier Medchemexpress LLC Supplier No. HY14654S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aspirin-d3 (acetylsalicylic acid-d3) is a deuterium-labeled analogue of aspirin provided for laboratory research. It is used as a stable-isotope internal standard or tracer in analytical, pharmacokinetic, and biochemical studies where a labeled form of aspirin is required. This compound is for research use only and is not for human consumption or clinical applications.

  • Molecular formula C9H5D3O4.
  • Molecular weight 183.18 g/mol.
  • High reported purity (~98.5%).
  • Soluble in DMSO for convenient sample preparation.
  • Supplied in small research pack sizes such as 5 mg for method development and standards.
  • Suitable as an internal standard for LC-MS and related assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.