Learn More
Medchemexpress LLC Acetylsalicylic acid-d3 | 921943-73-9 | MFCD09752106 | 98.5% | C9H5D3O4 | 10MG
SDP
Catalog No. 5000258485
Encompass_Preferred
Aspirin-d3 (acetylsalicylic acid-d3) is the deuterium-labeled analogue of aspirin used as a stable isotope internal standard in analytical and research workflows. It is provided in small research-scale quantities and is intended for use in quantitative mass spectrometry, method development, and pharmacokinetic or metabolic studies.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Chemical formula C9H5D3O4.
  • High purity suitable for analytical workflows (≈98.5%).
  • White to off-white solid, stable under standard storage conditions.
  • Available in small milligram quantities for method development.
Catalog No. 50-002-58485 Supplier Medchemexpress LLC Supplier No. HY14654S10MG
Aspirin-d3 (acetylsalicylic acid-d3) is the deuterium-labeled analogue of aspirin used as a stable isotope internal standard in analytical and research workflows. It is provided in small research-scale quantities and is intended for use in quantitative mass spectrometry, method development, and pharmacokinetic or metabolic studies.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Chemical formula C9H5D3O4.
  • High purity suitable for analytical workflows (≈98.5%).
  • White to off-white solid, stable under standard storage conditions.
  • Available in small milligram quantities for method development.