Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetylsalicylic acid-d4 | 97781-16-3 | MFCD06658866 | 97.7% | 184.18 g/mol | C9H4D4O4 | 5 MG
SDP

Catalog No. 5000222300
Encompass_Preferred

Aspirin-d4 (acetylsalicylic acid-d4) is a deuterium-labeled analogue of aspirin intended for use as an internal standard in quantitative GC-MS and LC-MS assays. Supplied in small vials, it provides a stable isotopic reference for analytical and pharmacokinetic studies requiring accurate measurement of aspirin levels.

  • Deuterium-labeled internal standard for aspirin quantification.
  • Suitable for GC-MS and LC-MS analytical methods.
  • High purity (97.7%) for reliable analytical performance.
  • Molecular formula C9H4D4O4; molecular weight 184.18 g/mol.
  • Available in small vial package sizes, including 5 mg.
  • CAS number 97781-16-3 for unambiguous identification.

Catalog No. 50-002-22300 Supplier Medchemexpress LLC Supplier No. HY14654S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aspirin-d4 (acetylsalicylic acid-d4) is a deuterium-labeled analogue of aspirin intended for use as an internal standard in quantitative GC-MS and LC-MS assays. Supplied in small vials, it provides a stable isotopic reference for analytical and pharmacokinetic studies requiring accurate measurement of aspirin levels.

  • Deuterium-labeled internal standard for aspirin quantification.
  • Suitable for GC-MS and LC-MS analytical methods.
  • High purity (97.7%) for reliable analytical performance.
  • Molecular formula C9H4D4O4; molecular weight 184.18 g/mol.
  • Available in small vial package sizes, including 5 mg.
  • CAS number 97781-16-3 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.