Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylsalicylic Acid, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S000498 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000498 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: Harmful if swallowed, causes skin irritation, causes serious eye irritation, may cause respiratory irritation. WARNING! : This product can expose you to Acetylsalicylic acid, which is known to the State of California to cause birth defects or other reproductive harm.
Catalog No. S000498 Supplier Aldon Corporation Supplier No. IS10027
Add to Cart
Add to Cart
Warning: Harmful if swallowed, causes skin irritation, causes serious eye irritation, may cause respiratory irritation. WARNING! : This product can expose you to Acetylsalicylic acid, which is known to the State of California to cause birth defects or other reproductive harm.

Chemical Identifiers

Molecular Weight (g/mol) 180.16 g/mol

Specifications

Quantity 100 g
Percent Purity > 99%
Grade Reagent
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.