Learn More
Medchemexpress LLC Acetylsalicylsalicylic acid | 530-75-6 | MFCD00143537 | 99.2% | 300.26 g/mol | C16H12O6 | 250 MG
SDP
Catalog No. 5000209457
Encompass_Preferred
Acetylsalicylsalicylic acid is an acetylated salicylic derivative provided for research use as a drug intermediate and immunogen. It is offered at high purity for experimental applications and is accompanied by application documents for safe laboratory handling.
  • Used as a drug intermediate and immunogen for research applications.
  • Reported to induce contact sensitivity, skin erythema, and slight edema in guinea pigs.
  • High purity (about 99.2%) with formula C16H12O6 and molecular weight 300.26 g/mol.
  • Available in small research pack sizes such as 250 MG for laboratory experiments.
  • Documentation available: data sheet, SDS, and COA for quality and safety information.
Catalog No. 50-002-09457 Supplier Medchemexpress LLC Supplier No. 250MGHYW087907
Acetylsalicylsalicylic acid is an acetylated salicylic derivative provided for research use as a drug intermediate and immunogen. It is offered at high purity for experimental applications and is accompanied by application documents for safe laboratory handling.
  • Used as a drug intermediate and immunogen for research applications.
  • Reported to induce contact sensitivity, skin erythema, and slight edema in guinea pigs.
  • High purity (about 99.2%) with formula C16H12O6 and molecular weight 300.26 g/mol.
  • Available in small research pack sizes such as 250 MG for laboratory experiments.
  • Documentation available: data sheet, SDS, and COA for quality and safety information.