Learn More
Cayman Chemical Acetylthiocholine (iodide), Purity: ≥95%, CAS: 1866-15-5, 50 g
SDP
Catalog No. 502950437
Encompass_Preferred
A substrate for AChE and an agonist of α4β2 subunit-containing nAChRs; binds to α4β2 subunit-containing nAChRs in SH-EP1 cells (IC50 = 7.1 µM); induces rubidium efflux in an agonist-induced rubidium efflux assay (EC50 = 100 µM); has been used as part of a thiocholine sensor for the detection of pesticides in complex sample types
Catalog No. 50-295-0437 Supplier Cayman Chemical Supplier No. 3431350 G
A substrate for AChE and an agonist of α4β2 subunit-containing nAChRs; binds to α4β2 subunit-containing nAChRs in SH-EP1 cells (IC50 = 7.1 µM); induces rubidium efflux in an agonist-induced rubidium efflux assay (EC50 = 100 µM); has been used as part of a thiocholine sensor for the detection of pesticides in complex sample types