Learn More
U.S. Pharmacopeia Acetyltriethyl Citrate, 77-89-4, MFCD00049378, 500mg
Catalog No. NC9408810
Encompass
Molecular Formula C14H22O8, Molecular Weight 318.320, Melting Point -43.6 °F (-42 °C), Boiling Point 566.6 °F (297 °C)
Catalog No. NC9408810 Supplier U.S. Pharmacopeia Supplier No. 1009923
Molecular Formula C14H22O8, Molecular Weight 318.320, Melting Point -43.6 °F (-42 °C), Boiling Point 566.6 °F (297 °C)