Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylurea 99.0+%, TCI America™
SDP

Catalog No. A012425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A012425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A012425G Supplier TCI America Supplier No. A012425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 591-07-1
Melting Point 215°C
Color White
Molecular Formula C3H6N2O2
Quantity 25 g
MDL Number MFCD00038140
Synonym acetylurea, acetamide, n-aminocarbonyl, n-acetylurea, acetylcarbamide, monoacetylurea, 1-acetylurea, acetic acid, ureide, n-aminocarbonyl acetamide, urea, acetyl, unii-8kru31i919
InChI Key GKRZNOGGALENQJ-UHFFFAOYSA-N
SMILES CC(=O)NC(=O)N
IUPAC Name N-carbamoylacetamide
Molecular Weight (g/mol) 102.093
PubChem CID 68956
Formula Weight 102.09
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Acetylurea
Show More Show Less
RTECSNumber AB4331500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.