Learn More
TARGETMOL CHEMICALS INC ACEVALTRATE 10MG
SDP
Catalog No. 502576183
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Acevaltrate (Acevaltratum) displays high cytotoxicity against GLC(4) a human small-cell lung cancer cell line and against COLO 320 a human colorectal cancer cell line with IC50 values of 1-6 uM. purity: 99%
Catalog No. 50-257-6183 Supplier TARGETMOL CHEMICALS INC Supplier No. TJP287210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Acevaltrate (Acevaltratum) displays high cytotoxicity against GLC(4) a human small-cell lung cancer cell line and against COLO 320 a human colorectal cancer cell line with IC50 values of 1-6 uM. purity: 99%