Learn More
Medchemexpress LLC HY-N2070 10mg Medchemexpress, Acevaltrate CAS:25161-41-5 Purity:>98%
SDP
Catalog No. 501933882
Encompass_Preferred
Medchemexpress, HY-N2070 10mg Acevaltrate CAS:25161-41-5 Acevaltrate, isolated from Valeriana glechomifolia, inhibits the Na + / K + -ATPase activity in the rat kidney and brain hemispheres with IC 50 s of 22.8±1.1 μM and 42.3±1.0 μM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3882 Supplier Medchemexpress LLC Supplier No. HYN207010MG
Medchemexpress, HY-N2070 10mg Acevaltrate CAS:25161-41-5 Acevaltrate, isolated from Valeriana glechomifolia, inhibits the Na + / K + -ATPase activity in the rat kidney and brain hemispheres with IC 50 s of 22.8±1.1 μM and 42.3±1.0 μM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.