Learn More
STEMCELL Technologies Animal Component-Free Cell Dissociation Kit, Size: 1 Kit
SDP
Catalog No. NC1457386
Encompass
Animal Component-Free Cell Dissociation Kit is optimized for dissociation and passaging of human stem and progenitor cells cultured in various media formulations. Animal Component-Free Cell Dissociation Kit includes ACF Enzymatic Dissociation Solution and ACF Enzyme Inhibition Solution. This kit was previously named MesenCult™-ACF Dissociation Kit. The name was changed to accommodate the use of this product with other cell types.
Catalog No. NC1457386 Supplier STEMCELL Technologies Supplier No. 05426
Animal Component-Free Cell Dissociation Kit is optimized for dissociation and passaging of human stem and progenitor cells cultured in various media formulations. Animal Component-Free Cell Dissociation Kit includes ACF Enzymatic Dissociation Solution and ACF Enzyme Inhibition Solution. This kit was previously named MesenCult™-ACF Dissociation Kit. The name was changed to accommodate the use of this product with other cell types.