Learn More
Aviva Systems Biology Corp ACHE Antibody - N-terminal region (ARP56761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACHE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACHE. Alternate names: ARACHE, N-ACHE, YT, ACEEApplicationIHC, WB, IF. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61978