Learn More
Medchemexpress LLC AChE-IN-27 | 177028-90-9 | 98.3% | 330.34 | 1 ML
SDP
Catalog No. 5000361623
Encompass_Preferred
AChE-IN-27 (compound 8c) is an AChE inhibitor with an IC50 of 0.19 μM. It is suitable for research in neurological diseases such as Alzheimer's disease, dementia, ataxia, and myasthenia gravis.
  • AChE inhibitor (IC50=0.19 μM)
  • Utilized in studies of neurological diseases
  • Applicable for Alzheimer's disease research
  • Used in studies for dementia, ataxia, and myasthenia gravis
Catalog No. 50-003-61623 Supplier Medchemexpress LLC Supplier No. HY14810810MM/1ML
AChE-IN-27 (compound 8c) is an AChE inhibitor with an IC50 of 0.19 μM. It is suitable for research in neurological diseases such as Alzheimer's disease, dementia, ataxia, and myasthenia gravis.
  • AChE inhibitor (IC50=0.19 μM)
  • Utilized in studies of neurological diseases
  • Applicable for Alzheimer's disease research
  • Used in studies for dementia, ataxia, and myasthenia gravis