Learn More
Medchemexpress LLC ACHE-IN-38 hydrochloride | 120013-39-0 | 325.84 | 10 G
SDP
Catalog No. 5000445869
Encompass_Preferred
ACHE-IN-38 hydrochloride inhibits the metabolic breakdown of the neurotransmitter acetylcholine (ACh) by the enzyme acetylcholinesterase (AChE). This action alleviates memory deficits in patients with Alzheimer's Disease by potentiating cholinergic transmission.
  • Inhibits acetylcholinesterase (AChE)
  • Alleviates memory deficits in Alzheimer's patients
  • Potentiates cholinergic transmission
Catalog No. 50-004-45869 Supplier Medchemexpress LLC Supplier No. HYW00357610G
ACHE-IN-38 hydrochloride inhibits the metabolic breakdown of the neurotransmitter acetylcholine (ACh) by the enzyme acetylcholinesterase (AChE). This action alleviates memory deficits in patients with Alzheimer's Disease by potentiating cholinergic transmission.
  • Inhibits acetylcholinesterase (AChE)
  • Alleviates memory deficits in Alzheimer's patients
  • Potentiates cholinergic transmission