Learn More
Medchemexpress LLC Ache-in-38 hydrochloride | 120013-39-0 | 325.84 | 100 G
SDP
Catalog No. 5000445640
Encompass_Preferred
ACHE-IN-38 hydrochloride (Compound 13b) is an inhibitor that targets cholinesterase (ChE) and neuronal signaling. It works by inhibiting the metabolic breakdown of the neurotransmitter acetylcholine (ACh) by the enzyme acetylcholinesterase (AChE). This action helps to alleviate memory deficits in patients with Alzheimer's Disease by potentiating cholinergic transmission.
  • Inhibits cholinesterase (ChE) and neuronal signaling
  • Alleviates memory deficits in Alzheimer's disease by potentiating cholinergic transmission
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year or -20°C for 6 months
Catalog No. 50-004-45640 Supplier Medchemexpress LLC Supplier No. HYW003576100G
ACHE-IN-38 hydrochloride (Compound 13b) is an inhibitor that targets cholinesterase (ChE) and neuronal signaling. It works by inhibiting the metabolic breakdown of the neurotransmitter acetylcholine (ACh) by the enzyme acetylcholinesterase (AChE). This action helps to alleviate memory deficits in patients with Alzheimer's Disease by potentiating cholinergic transmission.
  • Inhibits cholinesterase (ChE) and neuronal signaling
  • Alleviates memory deficits in Alzheimer's disease by potentiating cholinergic transmission
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year or -20°C for 6 months