Learn More
Medchemexpress LLC ACHE-IN-38 hydrochloride | 120013-39-0 | 325.84 | 50 G
SDP
Catalog No. 5000445409
Encompass_Preferred
ACHE-IN-38 hydrochloride (Standard) is the analytical standard of ACHE-IN-38 hydrochloride. This compound inhibits the metabolic breakdown of the neurotransmitter acetylcholine (ACh) by the enzyme acetylcholinesterase (AChE), which helps alleviate memory deficits in patients with Alzheimer's Disease by potentiating cholinergic transmission.
  • Analytical standard for research and analytical applications
  • Inhibits acetylcholinesterase (AChE)
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Alleviates memory deficits in Alzheimer's Disease patients
  • Potentiates cholinergic transmission
Catalog No. 50-004-45409 Supplier Medchemexpress LLC Supplier No. HYW00357650G
ACHE-IN-38 hydrochloride (Standard) is the analytical standard of ACHE-IN-38 hydrochloride. This compound inhibits the metabolic breakdown of the neurotransmitter acetylcholine (ACh) by the enzyme acetylcholinesterase (AChE), which helps alleviate memory deficits in patients with Alzheimer's Disease by potentiating cholinergic transmission.
  • Analytical standard for research and analytical applications
  • Inhibits acetylcholinesterase (AChE)
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Alleviates memory deficits in Alzheimer's Disease patients
  • Potentiates cholinergic transmission