Learn More
Cayman Chemical AChEeel PolycLNal ntIbod 300ug
SDP
Catalog No. 502995097
Encompass_Preferred
A potent inhibitor of E-selectin and ICAM-1 expression in HUVECs stimulated with TNF-a (IC50s 20 and 25 nM respectively)
Catalog No. 50-299-5097 Supplier Cayman Chemical Supplier No. 23856300 UG
A potent inhibitor of E-selectin and ICAM-1 expression in HUVECs stimulated with TNF-a (IC50s 20 and 25 nM respectively)