Specifications
| Certifications/Compliance | ANSI Z87.1 |
| Description | 8 base curve lens , wrap around design with side protection |
| Lens Tint | Clear |
| Frame Color | Gray |
| Temple Color | Clear with Gray Tips |
| Lens Material | Polycarbonate |