Learn More
NOTRAX™ Achilles™ Mat
GREENER_CHOICE
Catalog No. 19086032 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
3 x 60 ft.
Catalog No. Dimensions (L x W)
19-086-032 3 x 60 ft.
1 options
Catalog No. 19-086-032 Supplier NOTRAX™ Supplier No. 408R0336BL
  • $914.00
    $624.65 / Each of 1
    Save $289.35
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Achilles™ is an embossed ribbed top surface that enhances the mats slip resistance while providing increased worker fatigue relief
  • Ribbed textured embossed pattern
  • Basic anti-fatigue protection provided
  • PVC closed cell foam relieves leg and back discomfort
  • Provides a barrier to cold floors
  • Beveled edges on all four sides
Specifications
Dimensions (L x W) 3 x 60 ft.
Quantity 1 Ea.
Color Black