Learn More
Medchemexpress LLC Achn-975 tfa | 1410809-37-8 | 99.0% | 483.44 | 1 ML
SDP
Catalog No. 5000403272
Encompass_Preferred
ACHN-975 TFA is a selective LpxC inhibitor that demonstrates subnanomolar LpxC inhibitory activity. It is effective against a broad spectrum of gram-negative bacteria, exhibiting low MIC values (≤1 μg/mL).
  • Selective LpxC inhibitor.
  • Exhibits subnanomolar LpxC inhibitory activity.
  • Active against a wide range of gram-negative bacteria.
  • Demonstrates low MIC values (≤1 μg/mL).
Catalog No. 50-004-03272 Supplier Medchemexpress LLC Supplier No. HY19936A10MM/1ML
ACHN-975 TFA is a selective LpxC inhibitor that demonstrates subnanomolar LpxC inhibitory activity. It is effective against a broad spectrum of gram-negative bacteria, exhibiting low MIC values (≤1 μg/mL).
  • Selective LpxC inhibitor.
  • Exhibits subnanomolar LpxC inhibitory activity.
  • Active against a wide range of gram-negative bacteria.
  • Demonstrates low MIC values (≤1 μg/mL).