Learn More
ATCC Acholeplasma laidlawii
Catalog No. 502383547
Encompass
Acholeplasma laidlawii strain PG8 [NCTC 10116, PG8; A] is a whole-genome sequenced bacterial type strain isolated from sewage. This strain is recommended as a test microorganism for USP <63>.
Catalog No. 50-238-3547 Supplier ATCC Supplier No. 23206
Acholeplasma laidlawii strain PG8 [NCTC 10116, PG8; A] is a whole-genome sequenced bacterial type strain isolated from sewage. This strain is recommended as a test microorganism for USP <63>.