Learn More
ATCC Genomic DNA from Acholeplasma laidlawii strain PG8 (ATCC qCRM-23206D™)
Catalog No. 502382599
Encompass
Genomic DNA from Acholeplasma laidlawii strain PG8 (ATCC qCRM-23206D™) is a quantified certified reference material that supports USP General Chapter <63>. Produced under an ISO 17034-accredited process, it is used for inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-2599 Supplier ATCC Supplier No. 23206D
Genomic DNA from Acholeplasma laidlawii strain PG8 (ATCC qCRM-23206D™) is a quantified certified reference material that supports USP General Chapter <63>. Produced under an ISO 17034-accredited process, it is used for inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.