Learn More
ATCC Acholeplasma laidlawii; Quantitative CRM; genomic DNA
Catalog No. 502384041
Encompass
Quantified genomic DNA isolated from Acholeplasma laidlawii strain PG8 [NCTC 10116, PG8; A] (ATCC 23206). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-4041 Supplier ATCC Supplier No. QCRM23206D
Quantified genomic DNA isolated from Acholeplasma laidlawii strain PG8 [NCTC 10116, PG8; A] (ATCC 23206). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.