Learn More
ATCC Acholeplasma laidlawii; Strain PG8
Catalog No. 502383615
Encompass
Acholeplasma laidlawii strain PG8 [NCTC 10116, PG8; A] is a whole-genome sequenced bacterial type strain isolated from sewage. This titered product has applications in assay development, quality control and mycoplasma testing.
Catalog No. 50-238-3615 Supplier ATCC Supplier No. 23206TTR
Acholeplasma laidlawii strain PG8 [NCTC 10116, PG8; A] is a whole-genome sequenced bacterial type strain isolated from sewage. This titered product has applications in assay development, quality control and mycoplasma testing.