Learn More
Medchemexpress LLC HY-13060 5mg , ACHP (Hydrochloride) CAS:406209-26-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130605MG
Encompass_Preferred
HY-13060 5mg ACHP (Hydrochloride) CAS: 406209-26-5 ACHP Hydrochloride (IKK-2 Inhibitor VIII) is a highly potent and selective IKK-β inhibitor with an IC50 of 8.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0033