Learn More
Medchemexpress LLC Achra(97-116) | 663154-30-1 | 99.4% | 2252.56 | 1 MG
SDP
Catalog No. 5000397798
Encompass_Preferred
AChRα(97-116) is a synthetic peptide corresponding to the 97-116 region of the rat acetylcholine receptor α-subunit, and also an inducer of autoimmune diseases. It can be used in studies related to experimental autoimmune myasthenia gravis.
  • Does not induce significant proliferation of lymphocytes.
  • Maintains antigen-specific immune cell phenotype responsive to dopaminergic modulation.
  • Effectively detects elevated antibody levels in serum.
  • Stimulates T cells to differentiate into Tfh cells.
  • Stimulates lymph node-derived mononuclear cells to produce elevated percentages of pre-Tfh cells.
  • Induces experimental autoimmune myasthenia gravis in Lewis rats.
  • Drives elevated antibody production, reduced muscle clusters, and expansion of Tfh cells.
Catalog No. 50-003-97798 Supplier Medchemexpress LLC Supplier No. HYP57661MG
Add to Cart
Add to Cart
AChRα(97-116) is a synthetic peptide corresponding to the 97-116 region of the rat acetylcholine receptor α-subunit, and also an inducer of autoimmune diseases. It can be used in studies related to experimental autoimmune myasthenia gravis.
  • Does not induce significant proliferation of lymphocytes.
  • Maintains antigen-specific immune cell phenotype responsive to dopaminergic modulation.
  • Effectively detects elevated antibody levels in serum.
  • Stimulates T cells to differentiate into Tfh cells.
  • Stimulates lymph node-derived mononuclear cells to produce elevated percentages of pre-Tfh cells.
  • Induces experimental autoimmune myasthenia gravis in Lewis rats.
  • Drives elevated antibody production, reduced muscle clusters, and expansion of Tfh cells.