Learn More
Fisher Science Education™ Upright Compound Microscope Achromat Objective
SCI_ED
Catalog No. S72635 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
40X
Catalog No. Magnification Power
S72635 40X
1 options
Catalog No. S72635
Achromat Upright Microscope Objective with correction for color and spherical aberration.
  • RMS
  • DIN standard
  • Available in 4X or 40X
  • Achromat Microscope Objective
  • Correction for color and spherical aberration
Specifications
Type Achromat Objective
Autoclavable Not Autoclavable
Description Correction: 0.17, Threading: RMS, N.A.: 0.65
Length (Metric) 160 mm
Magnification Power 40X
Sterility Non-sterile
Voltage 0/1 V
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase