Learn More
Fisher Science Education™ Upright Compound Microscope Achromat Objective
SCI_ED
Catalog No. S72636 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
40X
4X
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
S72636 4X
S72635 40X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S72636
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Achromat Upright Microscope Objective with correction for color and spherical aberration.
  • RMS
  • DIN standard
  • Available in 4X or 40X
  • Achromat Microscope Objective
  • Correction for color and spherical aberration
Specifications
Type Achromat Objective
Autoclavable Not Autoclavable
Description Correction: 0.17, Threading: RMS, N.A.: 0.10
Length (Metric) 160 mm
Magnification Power 4X
Sterility Non-sterile
Voltage 0/1 V
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase