Learn More
Electron Microscopy Sciences Achromatic Magnifier, 2x
SDP
Supplier:  Electron Microscopy Sciences 681112X
Encompass_Preferred
The magnifying glass introduced here is the new version of the classic magnifying glass used by the famous detective Sherlock Holmes. This achromatic is more defined in its function and can give you more accurate details of the subject to be observed. This magnifier is composed by two optical glass lenses, which are cemented together. The image seen through this lens is free of color and gives sharp contours so that no vague lines will be found throughout the entire image field.
With a solid red wood handle and a red mark at the base of the lens frame, this reminds you to use the face having this red point upwards. Lens diameter is 37 mm.
Catalog No. 50-305-30