Learn More
ATCC Streptomyces albus (80614)
Catalog No. 502379983
Encompass
Produces salinomycin, salbostatin, and epi-17-deoxy (0-8)-salinomycin. This material is cited in a U.S. and/or other Patent and may not be used to infringe the patent claims.
Catalog No. 50-237-9983 Supplier ATCC Supplier No. 21838
Produces salinomycin, salbostatin, and epi-17-deoxy (0-8)-salinomycin. This material is cited in a U.S. and/or other Patent and may not be used to infringe the patent claims.