Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Achyranthoside D | 168009-91-4 | 1 MG
SDP

Catalog No. 5000417243
Encompass_Preferred

Achyranthoside D is a triterpene saponin derived from Achyranthes root. It is intended for research use only and has not been fully validated for medical applications.

  • Purity: 98.74%
  • Molecular weight: 1119.20
  • Molecular formula: C₅₃H₈₂O₂₅
  • Appearance: Solid
  • Color: White to off-white
  • Melting point: 205-206°C
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-17243 Supplier Medchemexpress LLC Supplier No. HYN79521MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Achyranthoside D is a triterpene saponin derived from Achyranthes root. It is intended for research use only and has not been fully validated for medical applications.

  • Purity: 98.74%
  • Molecular weight: 1119.20
  • Molecular formula: C₅₃H₈₂O₂₅
  • Appearance: Solid
  • Color: White to off-white
  • Melting point: 205-206°C
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.